missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-beta-phenyl-D-phenylalanine, 98%

Catalog No. AAH56885ME
Change view
Click to view available options
Quantity:
500 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH56885ME 500 mg
AAH5688503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH56885ME Supplier Thermo Scientific Chemicals Supplier No. H56885ME
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 143060-31-5 | C20H23NO4 | 341.41 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 143060-31-5
Molecular Formula C20H23NO4
Molecular Weight (g/mol) 341.41
MDL Number MFCD00191187
InChI Key TYJDOLCFYZSNQC-UHFFFAOYNA-N
Synonym boc-d-3,3-diphenylalanine, boc-d-ala 3,3-diphenyl-oh, r-2-tert-butoxycarbonyl amino-3,3-diphenylpropanoic acid, boc-3,3-diphenyl-d-alanine, r-n-boc-2-amino-3,3-diphenylpropionic acid, n-boc-beta-phenyl-d-phenylalanine, n-tert-butoxycarbonyl-beta-phenyl-d-phenylalanine, 2r-2-tert-butoxycarbonyl amino-3,3-diphenylpropanoic acid, n-tert-butoxycarbonyl-3-phenyl-l-phenylalanine, n-boc-b-phenyl-d-phenylalanine
PubChem CID 7019136
SMILES CC(C)(C)OC(=O)NC(C(C1=CC=CC=C1)C1=CC=CC=C1)C(O)=O

Specifications

Melting Point 157°C
Quantity 500 mg
Optical Rotation −30 (c=1 in Ethanol)
IUPAC Name 2-{[(tert-butoxy)carbonyl]amino}-3,3-diphenylpropanoic acid
Formula Weight 341.4
Percent Purity 98%
Chemical Name or Material N-Boc-beta-phenyl-D-phenylalanine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only