missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-beta-alanine, 99%

Catalog No. p-7026499
Change view
Click to view available options
Quantity:
2.5 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2252288 2.5 g
AAB2252209 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2252288 Supplier Thermo Scientific Chemicals Supplier No. B2252288

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3303-84-2 | C8H15NO4 | 189.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3303-84-2
Molecular Formula C8H15NO4
Molecular Weight (g/mol) 189.21
MDL Number MFCD00037291
InChI Key WCFJUSRQHZPVKY-UHFFFAOYSA-N
Synonym boc-beta-alanine, boc-beta-ala-oh, n-boc-beta-alanine, boc-b-ala-oh, boc-beta-ala, n-tert-butoxycarbonyl-beta-alanine, 3-tert-butoxycarbonyl amino propanoic acid, n-t-boc-beta-alanine, n-t-butyloxycarbonyl-beta-alanine, 3-tert-butoxycarbonylamino propanoic acid
PubChem CID 76809
IUPAC Name 3-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)NCCC(O)=O

Specifications

Melting Point 73°C to 76°C
Linear Formula (CH3)3CO2CNHCH2CH2CO2H
Quantity 2.5 g
Formula Weight 189.21
Percent Purity 99%
Chemical Name or Material N-Boc-beta-alanine
EINECSNumber 221-979-2
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only