missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-5-oxo-D-proline, 97%

Catalog No. aah5870103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5870103 1 g
AAH5870106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5870103 Supplier Thermo Scientific Chemicals Supplier No. H5870103
Only null left

CAS: 160347-90-0 | C10H15NO5 | 229.232 g/mol

Used as pharmaceutical intermediates and anesthetic agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as pharmaceutical intermediates and anesthetic agent.

Solubility
Slightly soluble in water.

Notes
Moisture Sensitive, store away from moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 160347-90-0
Molecular Formula C10H15NO5
Molecular Weight (g/mol) 229.232
MDL Number MFCD08458561
InChI Key MJLQPFJGZTYCMH-ZCFIWIBFSA-N
Synonym boc-d-pyr-oh, r-boc-5-oxopyrrolidine-2-carboxylic acid, r-1-tert-butoxycarbonyl-5-oxopyrrolidine-2-carboxylic acid, n-boc-d-pyroglutamic acid, 2r-1-tert-butoxy carbonyl-5-oxopyrrolidine-2-carboxylic acid, 2r-1-tert-butoxycarbonyl-5-oxopyrrolidine-2-carboxylic acid, boc-d-pglu-oh, ambotzbaa5600, n-boc-5-oxo-d-proline
PubChem CID 18645552
IUPAC Name (2R)-1-[(2-methylpropan-2-yl)oxycarbonyl]-5-oxopyrrolidine-2-carboxylic acid
SMILES CC(C)(C)OC(=O)N1C(CCC1=O)C(=O)O

Specifications

Melting Point 111°C to 116°C
Solubility Information Slightly soluble in water.
Quantity 1 g
Formula Weight 229.23
Optical Rotation +35° (c=1 in Chloroform)
Sensitivity Moisture sensitive
Percent Purity 97%
Chemical Name or Material N-Boc-5-oxo-D-proline
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only