accessibility menu, dialog, popup

UserName

CAS RN 160347-90-0

CH3H3CCH3OONOOOH

CAS RN 160347-90-0

IUPAC Name: 1-[(tert-butoxy)carbonyl]-5-oxopyrrolidine-2-carboxylic acid
Synonyms: 1-(tert-butoxycarbonyl)-5-oxopyrrolidine-2-carboxylic acid
Molecular Weight (g/mol): 229.23
Molecular Formula: C10H15NO5
InChi Key: MJLQPFJGZTYCMH-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)N1C(CCC1=O)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)