missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-4-oxo-L-proline methyl ester, 97%, Thermo Scientific Chemicals

Catalog No. p-7030226
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
250 mg
5 g
Catalog No. Quantity
AAH27782MD 250 mg
AAH2778206 5 g
2 options

Catalog No. AAH27782MD

Supplier: Thermo Scientific Chemicals H27782MD

Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 102195-80-2
Molecular Formula C11H17NO5
Molecular Weight (g/mol) 243.259
MDL Number MFCD01861778
InChI Key UPBHYYJZVWZCOZ-QMMMGPOBSA-N
Synonym boc-4-oxo-pro-ome, n-boc-4-oxo-l-proline methyl ester, s-1-tert-butyl 2-methyl 4-oxopyrrolidine-1,2-dicarboxylate, 1-tert-butyl 2-methyl 2s-4-oxopyrrolidine-1,2-dicarboxylate, 2s-1-boc-4-oxo-proline methyl ester, methyl 1-boc-4-oxo-proline, 1-boc-4-oxo-l-proline methyl ester, tert-butyl 2s-2-methoxycarbonyl-4-oxopyrrolidine-1-carboxylate, 2s-4-oxo-1-tert-butoxycarbonyl pyrrolidine-2-carboxylic acid methyl ester, 1,2-pyrrolidinedicarboxylic acid, 4-oxo-, 1-1,1-dimethylethyl 2-methyl ester, 2s
PubChem CID 10988485
IUPAC Name 1-O-tert-butyl 2-O-methyl (2S)-4-oxopyrrolidine-1,2-dicarboxylate
SMILES CC(C)(C)OC(=O)N1CC(=O)CC1C(=O)OC

Specifications

Melting Point 35°C to 40°C
Quantity 250 mg
Formula Weight 243.26
Percent Purity 97%
Chemical Name or Material N-Boc-4-oxo-L-proline methyl ester
Hazard Category H302+H312+H332-H318
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P363-P501c
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only