missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-4-nitro-L-phenylalanine, 95%

Catalog No. p-7034315
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5198603 1 g
AAH5198606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5198603 Supplier Thermo Scientific Chemicals Supplier No. H5198603
Only null left

CAS: 33305-77-0 | C14H18N2O6 | 310.306 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 33305-77-0
Molecular Formula C14H18N2O6
Molecular Weight (g/mol) 310.306
MDL Number MFCD00038128
InChI Key XBQADBXCNQPHHY-NSHDSACASA-N
Synonym boc-4-nitro-l-phenylalanine, boc-phe 4-no2-oh, boc-l-4-nitrophenylalanine, boc-l-4-nitrophe, n-boc-4-nitro-l-phenylalanine, s-2-tert-butoxycarbonyl amino-3-4-nitrophenyl propanoic acid, boc-l-4-no2-phe-oh, 2s-2-tert-butoxycarbonyl amino-3-4-nitrophenyl propanoic acid, n-tert-butoxycarbonyl-4-nitro-l-phenylalanine, 2s-2-tert-butoxy carbonyl amino-3-4-nitrophenyl propanoic acid
PubChem CID 7021882
SMILES CC(C)(C)OC(=O)NC(CC1=CC=C(C=C1)[N+](=O)[O-])C(=O)O

Specifications

Quantity 1 g
Beilstein 2820671
Optical Rotation +8° (c=1 in Methanol)
Formula Weight 310.31
Percent Purity 95%
Chemical Name or Material N-Boc-4-nitro-L-phenylalanine
EINECSNumber 251-450-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only