missing translation for 'onlineSavingsMsg'
Learn More

N-BOC-3-pyrrolidinone, 97%, Thermo Scientific™

$131.53 - $924.01

Chemical Identifiers

CAS 101385-93-7
Molecular Formula C9H15NO3
Molecular Weight (g/mol) 185.22
MDL Number MFCD01631194
InChI Key JSOMVCDXPUXKIC-UHFFFAOYSA-N
Synonym n-boc-3-pyrrolidinone, 1-boc-3-pyrrolidinone, 1-n-boc-3-pyrrolidinone, n-tert-butoxycarbonyl-3-pyrrolidinone, n-boc-pyrrolidine-3-one, boc-3-pyrrolidinone, 1-boc-3-pyrrolidone, 3-oxo-pyrrolidine-1-carboxylic acid tert-butyl ester, tert-butyl 3-oxopyrrolidinecarboxylate, 1-tert-butoxycarbonyl-3-pyrrolidone
PubChem CID 471360
IUPAC Name tert-butyl 3-oxopyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(=O)C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC397050010
View Documents Promotion Details
N/A 1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC397050100
View Documents Promotion Details
N/A 10g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

101385-93-7
185.22
JSOMVCDXPUXKIC-UHFFFAOYSA-N
471360
CC(C)(C)OC(=O)N1CCC(=O)C1
C9H15NO3
MFCD01631194
n-boc-3-pyrrolidinone, 1-boc-3-pyrrolidinone, 1-n-boc-3-pyrrolidinone, n-tert-butoxycarbonyl-3-pyrrolidinone, n-boc-pyrrolidine-3-one, boc-3-pyrrolidinone, 1-boc-3-pyrrolidone, 3-oxo-pyrrolidine-1-carboxylic acid tert-butyl ester, tert-butyl 3-oxopyrrolidinecarboxylate, 1-tert-butoxycarbonyl-3-pyrrolidone
tert-butyl 3-oxopyrrolidine-1-carboxylate

Specifications

101385-93-7
100.0
Authentic
C9H15NO3
n-boc-3-pyrrolidinone, 1-boc-3-pyrrolidinone, 1-n-boc-3-pyrrolidinone, n-tert-butoxycarbonyl-3-pyrrolidinone, n-boc-pyrrolidine-3-one, boc-3-pyrrolidinone, 1-boc-3-pyrrolidone, 3-oxo-pyrrolidine-1-carboxylic acid tert-butyl ester, tert-butyl 3-oxopyrrolidinecarboxylate, 1-tert-butoxycarbonyl-3-pyrrolidone
CC(C)(C)OC(=O)N1CCC(=O)C1
185.22
185.22
N-BOC-3-pyrrolidinone
97.5
>110°C
97.5% min. (GC)
MFCD01631194
JSOMVCDXPUXKIC-UHFFFAOYSA-N
tert-butyl 3-oxopyrrolidine-1-carboxylate
471360
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only