missing translation for 'onlineSavingsMsg'
Learn More

N-BOC-3-pyrrolidinone, 97%

Catalog No. AC397050010
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC397050010 1 g Glass bottle
AC397050100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC397050010 Supplier Thermo Scientific Chemicals Supplier No. 397050010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 101385-93-7 | C9H15NO3 | 185.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 101385-93-7
Molecular Formula C9H15NO3
Molecular Weight (g/mol) 185.22
MDL Number MFCD01631194
InChI Key JSOMVCDXPUXKIC-UHFFFAOYSA-N
Synonym n-boc-3-pyrrolidinone, 1-boc-3-pyrrolidinone, 1-n-boc-3-pyrrolidinone, n-tert-butoxycarbonyl-3-pyrrolidinone, n-boc-pyrrolidine-3-one, boc-3-pyrrolidinone, 1-boc-3-pyrrolidone, 3-oxo-pyrrolidine-1-carboxylic acid tert-butyl ester, tert-butyl 3-oxopyrrolidinecarboxylate, 1-tert-butoxycarbonyl-3-pyrrolidone
PubChem CID 471360
IUPAC Name tert-butyl 3-oxopyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(=O)C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 34°C to 38°C
Color Orange to White
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 185.22
Percent Purity 97%
Physical Form Low Melting Solid
Chemical Name or Material N-BOC-3-pyrrolidinone
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only