missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-3-iodo-L-alanine methyl ester, 98%

Catalog No. AAH6163803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6163803 1 g
AAH6163806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6163803 Supplier Thermo Scientific Chemicals Supplier No. H6163803
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 93267-04-0 | C9H16INO4 | 329.13 g/mol

N-Boc-3-iodo-L-alanine methyl ester is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-3-iodo-L-alanine methyl ester is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Soluble in water and 1% acetic acid.

Notes
Light sensitive. Store in dark. Store in cool, dry place in tightly closed container. Store away from oxidizing agent.

Chemical Identifiers

CAS 93267-04-0
Molecular Formula C9H16INO4
Molecular Weight (g/mol) 329.13
MDL Number MFCD00216579
InChI Key UGZBFCCHLUWCQI-LURJTMIESA-N
Synonym boc-beta-iodo-ala-ome, boc-3-iodo-l-alanine methyl ester, r-methyl 2-tert-butoxycarbonyl amino-3-iodopropanoate, n-tert-butoxycarbonyl-3-iodo-l-alanine methyl ester, l-n-boc-3-iodoalanine methyl ester, r-methyl 2-tert-butoxycarbonylamino-3-iodopropanoate, boc-3-iodo-l-ala-ome, n-boc-3-iodo-l-alanine methyl ester, r-boc-, a-iodo-ala-ome, methyl 2r-2-tert-butoxycarbonyl amino-3-iodopropanoate
PubChem CID 10903591
SMILES COC(=O)[C@H](CI)NC(=O)OC(C)(C)C

Specifications

Melting Point 50°C to 52°C
Color White
Beilstein 4422011
Solubility Information Soluble in water and 1% acetic acid.
IUPAC Name methyl (2R)-2-{[(tert-butoxy)carbonyl]amino}-3-iodopropanoate
Quantity 1 g
Formula Weight 329.13
Sensitivity Light sensitive
Percent Purity 98%
Physical Form Powder
Chemical Name or Material N-Boc-3-iodo-L-alanine methyl ester
Show More Show Less
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only