missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Boc-3-dimethylamino-DL-alanine, 97%
CAS: 851653-36-6 | C10H20N2O4 | 232.28 g/mol
$196.73 - $196.73
Chemical Identifiers
| CAS | 851653-36-6 |
|---|---|
| Molecular Formula | C10H20N2O4 |
| Molecular Weight (g/mol) | 232.28 |
| MDL Number | MFCD04972270 |
| InChI Key | VCDQZVYJKDSORW-UHFFFAOYSA-N |
| Synonym | 2-tert-butoxycarbonyl amino-3-dimethylamino propanoic acid, nalpha-boc-+/--2-amino-3-dimethylamino propionic acid, n-tert-butoxycarbonyl-3-dimethylamino alanine, n?-boc-2-amino-3-dimethylamino propionic acid, alanine,3-dimethylamino-n-1,1-dimethylethoxy c, 2-tert-butoxycarbonylamino-3-dimethylamino-propionic acid, 2-tert-butoxy carbonyl amino-3-dimethylamino propanoic acid, n-alpha-boc-+/--2-amino-3-dimethylamino propionic acid |
| PubChem CID | 13850981 |
| IUPAC Name | 3-(dimethylamino)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid |
| SMILES | CC(C)(C)OC(=O)NC(CN(C)C)C(=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH5241203
|
Thermo Scientific Chemicals
H5241203 |
1 g |
Each for $196.73
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
NotesStore in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Chemical Identifiers
| 851653-36-6 | |
| 232.28 | |
| VCDQZVYJKDSORW-UHFFFAOYSA-N | |
| 13850981 | |
| CC(C)(C)OC(=O)NC(CN(C)C)C(=O)O |
| C10H20N2O4 | |
| MFCD04972270 | |
| 2-tert-butoxycarbonyl amino-3-dimethylamino propanoic acid, nalpha-boc-+/--2-amino-3-dimethylamino propionic acid, n-tert-butoxycarbonyl-3-dimethylamino alanine, n?-boc-2-amino-3-dimethylamino propionic acid, alanine,3-dimethylamino-n-1,1-dimethylethoxy c, 2-tert-butoxycarbonylamino-3-dimethylamino-propionic acid, 2-tert-butoxy carbonyl amino-3-dimethylamino propanoic acid, n-alpha-boc-+/--2-amino-3-dimethylamino propionic acid | |
| 3-(dimethylamino)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid |
Specifications
| 851653-36-6 | |
| MFCD04972270 | |
| VCDQZVYJKDSORW-UHFFFAOYSA-N | |
| 3-(dimethylamino)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid | |
| 232.28 | |
| 232.28 | |
| N-Boc-3-dimethylamino-DL-alanine |
| C10H20N2O4 | |
| 2-tert-butoxycarbonyl amino-3-dimethylamino propanoic acid, nalpha-boc-+/--2-amino-3-dimethylamino propionic acid, n-tert-butoxycarbonyl-3-dimethylamino alanine, n?-boc-2-amino-3-dimethylamino propionic acid, alanine,3-dimethylamino-n-1,1-dimethylethoxy c, 2-tert-butoxycarbonylamino-3-dimethylamino-propionic acid, 2-tert-butoxy carbonyl amino-3-dimethylamino propanoic acid, n-alpha-boc-+/--2-amino-3-dimethylamino propionic acid | |
| CC(C)(C)OC(=O)NC(CN(C)C)C(=O)O | |
| 1 g | |
| 13850981 | |
| 97% |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only