accessibility menu, dialog, popup

UserName

CH3CH3H3COOHNNH3CCH3OOH

CAS RN 851653-36-6

IUPAC Name: 2-{[(tert-butoxy)carbonyl]amino}-3-(dimethylamino)propanoic acid
Synonyms: 2-[(tert-butoxycarbonyl)amino]-3-(dimethylamino)propanoic acid
Molecular Weight (g/mol): 232.28
Molecular Formula: C10H20N2O4
InChi Key: VCDQZVYJKDSORW-UHFFFAOYNA-N
SMILES: CN(C)CC(NC(=O)OC(C)(C)C)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)