Learn More
N-Boc-3-(4-pyridyl)-D-alanine, 95%
CAS: 37535-58-3 | C13H18N2O4 | 266.297 g/mol
$245.48 - $948.82
Chemical Identifiers
| CAS | 37535-58-3 |
|---|---|
| Molecular Formula | C13H18N2O4 |
| Molecular Weight (g/mol) | 266.297 |
| MDL Number | MFCD00672529 |
| InChI Key | FNYWDMKESUACOU-SNVBAGLBSA-N |
| Synonym | boc-d-4-pyridylalanine, boc-d-4-pal-oh, r-2-tert-butoxycarbonyl amino-3-pyridin-4-yl propanoic acid, boc-3-4-pyridyl-d-alanine, boc-d-4-pyridylala, boc-4'-pyridyl-d-ala, boc-3-4-pyridyl-d-ala-oh, n-tert-butoxycarbonyl-4-pyridyl-d-alanine, boc-d-3-4-pyridyl-alanine, r-2-tert-butoxycarbonylamino-3-pyridin-4-yl-propionic acid |
| PubChem CID | 2734503 |
| IUPAC Name | (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-pyridin-4-ylpropanoic acid |
| SMILES | CC(C)(C)OC(=O)NC(CC1=CC=NC=C1)C(=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6221503
|
Thermo Scientific Chemicals
H6221503 |
1 g |
Each for $245.48
|
|
|||||
|
AAH6221506
|
Thermo Scientific Chemicals
H6221506 |
5 g |
Each for $948.82
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 37535-58-3 | |
| 266.297 | |
| FNYWDMKESUACOU-SNVBAGLBSA-N | |
| 2734503 | |
| CC(C)(C)OC(=O)NC(CC1=CC=NC=C1)C(=O)O |
| C13H18N2O4 | |
| MFCD00672529 | |
| boc-d-4-pyridylalanine, boc-d-4-pal-oh, r-2-tert-butoxycarbonyl amino-3-pyridin-4-yl propanoic acid, boc-3-4-pyridyl-d-alanine, boc-d-4-pyridylala, boc-4'-pyridyl-d-ala, boc-3-4-pyridyl-d-ala-oh, n-tert-butoxycarbonyl-4-pyridyl-d-alanine, boc-d-3-4-pyridyl-alanine, r-2-tert-butoxycarbonylamino-3-pyridin-4-yl-propionic acid | |
| (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-pyridin-4-ylpropanoic acid |
Specifications
| 223°C to 229°C | |
| White | |
| MFCD00672529 | |
| boc-d-4-pyridylalanine, boc-d-4-pal-oh, r-2-tert-butoxycarbonyl amino-3-pyridin-4-yl propanoic acid, boc-3-4-pyridyl-d-alanine, boc-d-4-pyridylala, boc-4'-pyridyl-d-ala, boc-3-4-pyridyl-d-ala-oh, n-tert-butoxycarbonyl-4-pyridyl-d-alanine, boc-d-3-4-pyridyl-alanine, r-2-tert-butoxycarbonylamino-3-pyridin-4-yl-propionic acid | |
| CC(C)(C)OC(=O)NC(CC1=CC=NC=C1)C(=O)O | |
| 1 g | |
| 2734503 | |
| Air and moisture sensitive | |
| Powder |
| 37535-58-3 | |
| C13H18N2O4 | |
| 4195478 | |
| FNYWDMKESUACOU-SNVBAGLBSA-N | |
| (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-pyridin-4-ylpropanoic acid | |
| 266.297 | |
| 266.29 | |
| 95% | |
| N-Boc-3-(4-pyridyl)-D-alanine |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold; Store under Argon
RUO – Research Use Only