missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-3-(4-pyridyl)-D-alanine, 95%

Catalog No. aah6221506
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6221506 5 g
AAH6221503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6221506 Supplier Thermo Scientific Chemicals Supplier No. H6221506
Only null left

CAS: 37535-58-3 | C13H18N2O4 | 266.297 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 37535-58-3
Molecular Formula C13H18N2O4
Molecular Weight (g/mol) 266.297
MDL Number MFCD00672529
InChI Key FNYWDMKESUACOU-SNVBAGLBSA-N
Synonym boc-d-4-pyridylalanine, boc-d-4-pal-oh, r-2-tert-butoxycarbonyl amino-3-pyridin-4-yl propanoic acid, boc-3-4-pyridyl-d-alanine, boc-d-4-pyridylala, boc-4'-pyridyl-d-ala, boc-3-4-pyridyl-d-ala-oh, n-tert-butoxycarbonyl-4-pyridyl-d-alanine, boc-d-3-4-pyridyl-alanine, r-2-tert-butoxycarbonylamino-3-pyridin-4-yl-propionic acid
PubChem CID 2734503
IUPAC Name (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-pyridin-4-ylpropanoic acid
SMILES CC(C)(C)OC(=O)NC(CC1=CC=NC=C1)C(=O)O

Specifications

Melting Point 223°C to 229°C
Color White
Beilstein 4195478
Quantity 5 g
Formula Weight 266.29
Sensitivity Air and moisture sensitive
Percent Purity 95%
Physical Form Powder
Chemical Name or Material N-Boc-3-(4-pyridyl)-D-alanine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only