missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-3-(4-biphenylyl)-D-alanine, 95%, Thermo Scientific Chemicals

Catalog No. AAH6349406
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6349406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6349406 Supplier Thermo Scientific Chemicals Supplier No. H6349406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 128779-47-5 | Molecular Formula: C20H23NO4 | Molecular Weight: 341.41 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 128779-47-5
Molecular Formula C20H23NO4
Molecular Weight (g/mol) 341.41
MDL Number MFCD00191185
InChI Key NBVVKAUSAGHTSU-UHFFFAOYNA-N
Synonym boc-d-4,4'-biphenylalanine, boc-d-4,4-biphenylalanine, boc-3-4-biphenylyl-d-alanine, r-3-1,1'-biphenyl-4-yl-2-tert-butoxycarbonyl amino propanoic acid, boc-d-bip-oh, boc-d-4-phenylphenylalanine, boc-4-biphenyl-d-ala, boc-4-phenyl-d-phe-oh, boc-d-bph-oh, boc-d-bip 4,4'-oh
PubChem CID 7019133
SMILES CC(C)(C)OC(=O)NC(CC1=CC=C(C=C1)C1=CC=CC=C1)C(O)=O

Specifications

IUPAC Name 3-{[1,1'-biphenyl]-4-yl}-2-{[(tert-butoxy)carbonyl]amino}propanoic acid
Quantity 5 g
Formula Weight 341.41
Percent Purity 95%
Physical Form Solid
Chemical Name or Material N-Boc-3-(4-biphenylyl)-D-alanine
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only