accessibility menu, dialog, popup

UserName

H3CH3CCH3OONHOHO

CAS RN 128779-47-5

IUPAC Name: 3-{[1,1'-biphenyl]-4-yl}-2-{[(tert-butoxy)carbonyl]amino}propanoic acid
Synonyms: 3-{[1,1'-biphenyl]-4-yl}-2-[(tert-butoxycarbonyl)amino]propanoic acid
Molecular Weight (g/mol): 341.41
Molecular Formula: C20H23NO4
InChi Key: NBVVKAUSAGHTSU-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC(CC1=CC=C(C=C1)C1=CC=CC=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)