missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-3,3-diphenyl-L-alanine, 95%

Catalog No. p-7034380
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5206303 1 g
AAH5206306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5206303 Supplier Thermo Scientific Chemicals Supplier No. H5206303
Only null left

CAS: 138662-63-2 | C20H23NO4 | 341.407 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 138662-63-2
Molecular Formula C20H23NO4
Molecular Weight (g/mol) 341.407
MDL Number MFCD00191186
InChI Key TYJDOLCFYZSNQC-KRWDZBQOSA-N
Synonym boc-l-3,3-diphenylalanine, boc-3,3-diphenyl-l-alanine, s-2-tert-butoxycarbonyl amino-3,3-diphenylpropanoic acid, s-n-boc-2-amino-3,3-diphenylpropionic acid, n-boc-beta-phenyl-l-phenylalanine, boc-beta-phenyl-phe-oh, n-tert-butoxycarbonyl-beta-phenyl-l-phenylalanine, s-2-tert-butoxycarbonylamino-3,3-diphenylpropanoic acid, 2s-2-tert-butoxycarbonyl amino-3,3-diphenylpropanoic acid, pubchem20362
PubChem CID 2761487
IUPAC Name (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3,3-diphenylpropanoic acid
SMILES CC(C)(C)OC(=O)NC(C(C1=CC=CC=C1)C2=CC=CC=C2)C(=O)O

Specifications

Melting Point 160°C (decomposition)
Quantity 1 g
Beilstein 4880641
Formula Weight 341.41
Percent Purity 95%
Chemical Name or Material N-Boc-3,3-diphenyl-L-alanine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only