missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-1H-pyrazole-1-carboxamidine, 98+%

Catalog No. p-7036145
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5439406 5 g
AAH5439414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5439406 Supplier Thermo Scientific Chemicals Supplier No. H5439406
Only null left

CAS: 152120-61-1 | C9H14N4O2 | 210.237 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 152120-61-1
Molecular Formula C9H14N4O2
Molecular Weight (g/mol) 210.237
MDL Number MFCD00216663
InChI Key IGSFMHYSWZUENI-UHFFFAOYSA-N
Synonym n-boc-1-guanylpyrazole, n-boc-1h-pyrazole-1-carboxamidine, n-boc-pyrazole-1-carboxamidine, n-tert-butoxycarbonyl-1h-pyrazole-1-carboxamidine, tert-butyl 1e-amino 1h-pyrazol-1-yl methylene carbamate, n-tert-butoxycarbonyl-1 h-pyrazole-1-carboxamidine, h-pyrazole-1-carboxamidine hcl, 1h-pyrazole-n-t-butyloxycarbonyl-1-carboxamidine, tert-butoxy-n-iminopyrazolylmethyl carboxamide, tert-butyl imino 1h-pyrazol-1-yl methylcarbamate
PubChem CID 9605068
IUPAC Name tert-butyl (NE)-N-[amino(pyrazol-1-yl)methylidene]carbamate
SMILES CC(C)(C)OC(=O)N=C(N)N1C=CC=N1

Specifications

Melting Point 97°C to 101°C
Quantity 5 g
Beilstein 6141956
Formula Weight 210.23
Percent Purity ≥98%
Chemical Name or Material N-Boc-1H-pyrazole-1-carboxamidine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only