accessibility menu, dialog, popup

UserName

CAS RN 152120-61-1

IUPAC Name: tert-butyl N-[amino(1H-pyrazol-1-yl)methylidene]carbamate
Synonyms: tert-butyl N-[amino(pyrazol-1-yl)methylidene]carbamate
Molecular Weight (g/mol): 210.24
Molecular Formula: C9H14N4O2
InChi Key: IGSFMHYSWZUENI-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N=C(N)N1C=CC=N1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)