missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-1,4-diaminobutane, 97+%

CAS: 68076-36-8 | C9H20N2O2 | 188.271 g/mol

Supplier:  Thermo Scientific Chemicals H56499AA

 View more versions of this product

Catalog No. AAH56499AA


Only null left

Description

Description

N-Boc-1,4-diaminobutane is used in preparation of pharmacologically active compounds, preparation of spermidine analogues and in introduction of a C4-spacer. It serve as intermediates of medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-1,4-diaminobutane is used in preparation of pharmacologically active compounds, preparation of spermidine analogues and in introduction of a C4-spacer. It serve as intermediates of medicine.

Notes
Incompatible with oxidizing agents. Store at room temperature.
Specifications

Specifications

N-Boc-1,4-diaminobutane
C9H20N2O2
(CH3)3COCONH(CH2)4NH2
UN2735
tert-butyl n-4-aminobutyl carbamate, n-boc-1,4-butanediamine, n-boc-1,4-diaminobutane, tert-butyl 4-aminobutyl carbamate, tert-butyl 4-aminobutylcarbamate, boc-1,4-diaminobutane, n-boc-1,4-butandiamine, n-4-aminobutyl carbamic acid tert-butyl ester, n-tert-butoxycarbonyl-1,4-diaminobutane, mono-4-n-boc-1,4-diaminobutane
0.984
tert-butyl N-(4-aminobutyl)carbamate
5 mL
4351
98%
68076-36-8
1.46
MFCD00210019
1937878
ZFQWJXFJJZUVPI-UHFFFAOYSA-N
CC(C)(C)OC(=O)NCCCCN
188.271
109°C (228°F)
188.27

Chemical Identifiers

68076-36-8
188.271
ZFQWJXFJJZUVPI-UHFFFAOYSA-N
4351
CC(C)(C)OC(=O)NCCCCN
C9H20N2O2
MFCD00210019
tert-butyl n-4-aminobutyl carbamate, n-boc-1,4-butanediamine, n-boc-1,4-diaminobutane, tert-butyl 4-aminobutyl carbamate, tert-butyl 4-aminobutylcarbamate, boc-1,4-diaminobutane, n-boc-1,4-butandiamine, n-4-aminobutyl carbamic acid tert-butyl ester, n-tert-butoxycarbonyl-1,4-diaminobutane, mono-4-n-boc-1,4-diaminobutane
tert-butyl N-(4-aminobutyl)carbamate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only