missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-1,4-diaminobutane, 97+%

Catalog No. p-7037704
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH56499XF 1 mL
AAH56499AA 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH56499XF Supplier Thermo Scientific Chemicals Supplier No. H56499XF

CAS: 68076-36-8 | C9H20N2O2 | 188.271 g/mol

N-Boc-1,4-diaminobutane is used in preparation of pharmacologically active compounds, preparation of spermidine analogues and in introduction of a C4-spacer. It serve as intermediates of medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-1,4-diaminobutane is used in preparation of pharmacologically active compounds, preparation of spermidine analogues and in introduction of a C4-spacer. It serve as intermediates of medicine.

Notes
Incompatible with oxidizing agents. Store at room temperature.

Chemical Identifiers

CAS 68076-36-8
Molecular Formula C9H20N2O2
Molecular Weight (g/mol) 188.271
MDL Number MFCD00210019
InChI Key ZFQWJXFJJZUVPI-UHFFFAOYSA-N
Synonym tert-butyl n-4-aminobutyl carbamate, n-boc-1,4-butanediamine, n-boc-1,4-diaminobutane, tert-butyl 4-aminobutyl carbamate, tert-butyl 4-aminobutylcarbamate, boc-1,4-diaminobutane, n-boc-1,4-butandiamine, n-4-aminobutyl carbamic acid tert-butyl ester, n-tert-butoxycarbonyl-1,4-diaminobutane, mono-4-n-boc-1,4-diaminobutane
PubChem CID 4351
IUPAC Name tert-butyl N-(4-aminobutyl)carbamate
SMILES CC(C)(C)OC(=O)NCCCCN

Specifications

Refractive Index 1.46
Linear Formula (CH3)3COCONH(CH2)4NH2
UN Number UN2735
Beilstein 1937878
Density 0.984
Quantity 1 mL
Flash Point 109°C (228°F)
Formula Weight 188.27
Percent Purity 98%
Chemical Name or Material N-Boc-1,4-diaminobutane
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only