missing translation for 'onlineSavingsMsg'
Learn More

N-BOC-1,3-diaminopropane, 97%

Catalog No. p-4258785
Change view
Click to view available options
Quantity:
5 mL
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC442730050 5 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC442730050 Supplier Thermo Scientific Chemicals Supplier No. 442730050
Only null left

CAS: 75178-96-0 | C8H19N2O2 | 175.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 75178-96-0
Molecular Formula C8H19N2O2
Molecular Weight (g/mol) 175.25
MDL Number MFCD00210021
InChI Key POHWAQLZBIMPRN-UHFFFAOYSA-O
Synonym n-boc-1,3-propanediamine, n-boc-1,3-diaminopropane, tert-butyl n-3-aminopropyl carbamate, tert-butyl 3-aminopropyl carbamate, n-3-aminopropyl carbamic acid tert-butyl ester, tert-butyl 3-aminopropylcarbamate, 1-boc-amino-1,3-propanediamine, 3-amino-propyl-carbamic acid tert-butyl ester, n-t-boc-1,3-propanediamine, n-tert-butoxycarbonyl-1,3-diaminopropane
PubChem CID 2735700
IUPAC Name tert-butyl N-(3-aminopropyl)carbamate
SMILES CC(C)(C)OC(=O)NCCC[NH3+]

Specifications

Melting Point 22.0°C
Boiling Point 94.0°C to 96.0°C (0.2 Torr)
CAS Min % 96.0
CAS Max % 100.0
Color Colorless
Density 0.9980g/mL
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Linear Formula (CH3)3CHOC(O)NH(CH2)3NH2
Quantity 5 mL
Specific Gravity 0.998
Formula Weight 174.24
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material N-BOC-1, 3-diaminopropane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only