missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-1,3-diaminopropane, 95%

Catalog No. AAH5030403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5030403 1 g
AAH5030406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5030403 Supplier Thermo Scientific Chemicals Supplier No. H5030403
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 75178-96-0 | C8H19N2O2 | 175.25 g/mol

N-Boc-1,3-diaminopropane is used in the preparation of spermidine analogues as well as in the preparation of pharmacologically active compounds. It is also used in suzuki reaction. Further, it is used in the preparation of [3-(3-cyano-propylamino)-propyl]-carbamic acid tert-butyl ester.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-1,3-diaminopropane is used in the preparation of spermidine analogues as well as in the preparation of pharmacologically active compounds. It is also used in suzuki reaction. Further, it is used in the preparation of [3-(3-cyano-propylamino)-propyl]-carbamic acid tert-butyl ester.

Solubility
Miscible with methanol.

Notes
Hygroscopic. Air sensitive. Incompatible with oxidizing agents and acids.

Chemical Identifiers

CAS 75178-96-0
Molecular Formula C8H19N2O2
Molecular Weight (g/mol) 175.25
MDL Number MFCD00210021
InChI Key POHWAQLZBIMPRN-UHFFFAOYSA-O
Synonym n-boc-1,3-propanediamine, n-boc-1,3-diaminopropane, tert-butyl n-3-aminopropyl carbamate, tert-butyl 3-aminopropyl carbamate, n-3-aminopropyl carbamic acid tert-butyl ester, tert-butyl 3-aminopropylcarbamate, 1-boc-amino-1,3-propanediamine, 3-amino-propyl-carbamic acid tert-butyl ester, n-t-boc-1,3-propanediamine, n-tert-butoxycarbonyl-1,3-diaminopropane
PubChem CID 2735700
SMILES CC(C)(C)OC(=O)NCCC[NH3+]

Specifications

Melting Point ∼22°C
Linear Formula (CH3)3COCONHCH2CH2CH2NH2
UN Number UN3259
Solubility Information Miscible with methanol.
IUPAC Name tert-butyl N-(3-azaniumylpropyl)carbamate
Quantity 1 g
Formula Weight 174.24
Sensitivity Air Sensitive
Percent Purity 95%
Chemical Name or Material N-Boc-1,3-diaminopropane
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only