missing translation for 'onlineSavingsMsg'
Learn More

N-[Bis(4-methoxyphenyl)methylene]benzylamine 98.0+%, TCI America™

Catalog No. B01371G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B01371G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B01371G Supplier TCI America Supplier No. B01371G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 524-96-9
Molecular Formula C22H21NO2
Molecular Weight (g/mol) 331.415
MDL Number MFCD00025824
InChI Key IZZONQFQTGCWNC-UHFFFAOYSA-N
Synonym N-(4,4′C-Dimethoxybenzhydrylidene)benzylamine, Schoenberg′Cs Reagent
PubChem CID 68233
IUPAC Name N-benzyl-1,1-bis(4-methoxyphenyl)methanimine
SMILES COC1=CC=C(C=C1)C(=NCC2=CC=CC=C2)C3=CC=C(C=C3)OC

Specifications

Melting Point 91°C
Color Yellow
Quantity 1 g
Formula Weight 331.42
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-[Bis(4-methoxyphenyl)methylene]benzylamine