missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N″-Biotinyl-6-aminohexanoylhydrazide 97.0+%, TCI America™
Supplier: TCI America H107125MG
Specifications
| N′′-Biotinyl-6-aminohexanoylhydrazide | |
| 211°C | |
| C16H29N5O3S | |
| 25 mg | |
| IJJWOSAXNHWBPR-HUBLWGQQSA-N | |
| 5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]-N-(6-hydrazinyl-6-oxohexyl)pentanamide | |
| 83900 | |
| ≥97.0% (HPLC) |
| 109276-34-8 | |
| White | |
| MFCD00077659 | |
| Biotin-LC-Hydrazide, N-(6-Hydrazinyl-6-oxohexyl)biotinamide | |
| C1C2C(C(S1)CCCCC(=O)NCCCCCC(=O)NN)NC(=O)N2 | |
| 371.5 | |
| 371.50 | |
| Crystalline Powder |
Chemical Identifiers
| 109276-34-8 | |
| 371.5 | |
| IJJWOSAXNHWBPR-HUBLWGQQSA-N | |
| 83900 | |
| C1C2C(C(S1)CCCCC(=O)NCCCCCC(=O)NN)NC(=O)N2 |
| C16H29N5O3S | |
| MFCD00077659 | |
| Biotin-LC-Hydrazide, N-(6-Hydrazinyl-6-oxohexyl)biotinamide | |
| 5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]-N-(6-hydrazinyl-6-oxohexyl)pentanamide |
Safety and Handling
TSCA : No
Recommended Storage : Freezer (-20 °C)