missing translation for 'onlineSavingsMsg'
Learn More

N″-Biotinyl-6-aminohexanoylhydrazide 97.0+%, TCI America™

Catalog No. p-7130890
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H107125MG 25 mg
H1071100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H107125MG Supplier TCI America Supplier No. H107125MG
Only null left

Chemical Identifiers

CAS 109276-34-8
Molecular Formula C16H29N5O3S
Molecular Weight (g/mol) 371.5
MDL Number MFCD00077659
InChI Key IJJWOSAXNHWBPR-HUBLWGQQSA-N
Synonym Biotin-LC-Hydrazide, N-(6-Hydrazinyl-6-oxohexyl)biotinamide
PubChem CID 83900
IUPAC Name 5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]-N-(6-hydrazinyl-6-oxohexyl)pentanamide
SMILES C1C2C(C(S1)CCCCC(=O)NCCCCCC(=O)NN)NC(=O)N2

Specifications

Melting Point 211°C
Color White
Quantity 25 mg
Formula Weight 371.50
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N′′-Biotinyl-6-aminohexanoylhydrazide
TSCA No
Recommended Storage Freezer (-20 °C)