Learn More
N-Benzylthiourea, 98%
CAS: 621-83-0 | C8H10N2S | 166.24 g/mol
Supplier: Thermo Scientific Chemicals L0940014
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| N-Benzylthiourea | |
| 162°C to 166°C | |
| C8H10N2S | |
| 25 g | |
| 972372 | |
| UCGFRIAOVLXVKL-UHFFFAOYSA-N | |
| benzylthiourea | |
| 737375 | |
| 98% |
| 621-83-0 | |
| Odorless | |
| MFCD00041370 | |
| UN2811 | |
| 1-benzylthiourea, 1-benzyl-2-thiourea, n-benzylthiourea, thiourea, phenylmethyl, urea, 1-benzyl-2-thio, unii-mrp6y7412k, amino benzylamino methane-1-thione, n-benzylthioformamide, benzylthioharnstoff, n-benzyl thiourea | |
| NC(=S)NCC1=CC=CC=C1 | |
| 166.24 | |
| 166.25 |
Chemical Identifiers
| 621-83-0 | |
| 166.24 | |
| UCGFRIAOVLXVKL-UHFFFAOYSA-N | |
| 737375 | |
| NC(=S)NCC1=CC=CC=C1 |
| C8H10N2S | |
| MFCD00041370 | |
| 1-benzylthiourea, 1-benzyl-2-thiourea, n-benzylthiourea, thiourea, phenylmethyl, urea, 1-benzyl-2-thio, unii-mrp6y7412k, amino benzylamino methane-1-thione, n-benzylthioformamide, benzylthioharnstoff, n-benzyl thiourea | |
| benzylthiourea |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P264b-P270-P301+P310-P330-P501c
H301
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 210-709-9
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only