missing translation for 'onlineSavingsMsg'
Learn More

N-Benzylthiourea, 98%

Catalog No. AAL0940022
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0940022 100 g
AAL0940006 5 g
AAL0940014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0940022 Supplier Thermo Scientific Chemicals Supplier No. L0940022
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 621-83-0 | C8H10N2S | 166.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 621-83-0
Molecular Formula C8H10N2S
Molecular Weight (g/mol) 166.24
MDL Number MFCD00041370
InChI Key UCGFRIAOVLXVKL-UHFFFAOYSA-N
Synonym 1-benzylthiourea, 1-benzyl-2-thiourea, n-benzylthiourea, thiourea, phenylmethyl, urea, 1-benzyl-2-thio, unii-mrp6y7412k, amino benzylamino methane-1-thione, n-benzylthioformamide, benzylthioharnstoff, n-benzyl thiourea
PubChem CID 737375
IUPAC Name benzylthiourea
SMILES NC(=S)NCC1=CC=CC=C1

Specifications

Melting Point 162°C to 166°C
Odor Odorless
Quantity 100 g
UN Number UN2811
Beilstein 972372
Formula Weight 166.25
Percent Purity 98%
Chemical Name or Material N-Benzylthiourea
Hazard Category H301
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 210-709-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only