missing translation for 'onlineSavingsMsg'
Learn More

N-(Benzyloxycarbonyloxy)succinimide, 98%

Catalog No. p-7081951
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1215314 25 g
AAA1215322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1215314 Supplier Thermo Scientific Chemicals Supplier No. A1215314
Only null left

CAS: 13139-17-8 | C12H11NO5 | 249.222 g/mol

N-(N-Benzyloxycarbonyloxy)succinimide is a reagent used in the preparation of peptide thioacids view peptide precursors. Also used in the preparation of amino glycoside derivatives for the function of topoisomerase for both human and bacterial inhibition.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-(N-Benzyloxycarbonyloxy)succinimide is a reagent used in the preparation of peptide thioacids view peptide precursors. Also used in the preparation of amino glycoside derivatives for the function of topoisomerase for both human and bacterial inhibition.

Solubility
Soluble in chloroform, methanol, acetone (0.1 g/mL).

Notes
Moisture sensitive. Storage temperature 2-8°C. Store away from oxidizing agents, water/ moisture.

Chemical Identifiers

CAS 13139-17-8
Molecular Formula C12H11NO5
Molecular Weight (g/mol) 249.222
MDL Number MFCD00005513
InChI Key MJSHDCCLFGOEIK-UHFFFAOYSA-N
Synonym n-benzyloxycarbonyloxy succinimide, cbz-osu, cbz-onsu, benzyl 2,5-dioxopyrrolidin-1-yl carbonate, benzyl succinimido carbonate, z-osu, n-carbobenzoxyoxysuccinimide, n-benzyloxycarbonyloxy-succinimide, cbzosu, carbobenzoxy succinimide
PubChem CID 83172
IUPAC Name benzyl (2,5-dioxopyrrolidin-1-yl) carbonate
SMILES C1CC(=O)N(C1=O)OC(=O)OCC2=CC=CC=C2

Specifications

Melting Point 77°C to 82°C
Quantity 25 g
Beilstein 1387927
Sensitivity Moisture sensitive
Solubility Information Soluble in chloroform,methanol,acetone (0.1g/mL).
Formula Weight 249.23
Percent Purity 99%
Chemical Name or Material N- (Benzyloxycarbonyloxy)succinimide
EINECSNumber 236-075-3
TSCA No
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only