missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(Benzyloxycarbonyloxy)succinimide, 98%
CAS: 13139-17-8 | C12H11NO5 | 249.22 g/mol
$306.41 - $306.41
Chemical Identifiers
| CAS | 13139-17-8 |
|---|---|
| Molecular Formula | C12H11NO5 |
| Molecular Weight (g/mol) | 249.22 |
| MDL Number | MFCD00005513 |
| InChI Key | MJSHDCCLFGOEIK-UHFFFAOYSA-N |
| Synonym | n-benzyloxycarbonyloxy succinimide, cbz-osu, cbz-onsu, benzyl 2,5-dioxopyrrolidin-1-yl carbonate, benzyl succinimido carbonate, z-osu, n-carbobenzoxyoxysuccinimide, n-benzyloxycarbonyloxy-succinimide, cbzosu, carbobenzoxy succinimide |
| PubChem CID | 83172 |
| IUPAC Name | benzyl (2,5-dioxopyrrolidin-1-yl) carbonate |
| SMILES | C1CC(=O)N(C1=O)OC(=O)OCC2=CC=CC=C2 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC211171000
|
Thermo Scientific Chemicals
211171000 |
100 g | Glass bottle |
Each for $306.41
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 13139-17-8 | |
| 249.22 | |
| MJSHDCCLFGOEIK-UHFFFAOYSA-N | |
| 83172 | |
| C1CC(=O)N(C1=O)OC(=O)OCC2=CC=CC=C2 |
| C12H11NO5 | |
| MFCD00005513 | |
| n-benzyloxycarbonyloxy succinimide, cbz-osu, cbz-onsu, benzyl 2,5-dioxopyrrolidin-1-yl carbonate, benzyl succinimido carbonate, z-osu, n-carbobenzoxyoxysuccinimide, n-benzyloxycarbonyloxy-succinimide, cbzosu, carbobenzoxy succinimide | |
| benzyl (2,5-dioxopyrrolidin-1-yl) carbonate |
Specifications
| 13139-17-8 | |
| 100.0 | |
| White | |
| Authentic | |
| Glass bottle | |
| MFCD00005513 | |
| n-benzyloxycarbonyloxy succinimide, cbz-osu, cbz-onsu, benzyl 2,5-dioxopyrrolidin-1-yl carbonate, benzyl succinimido carbonate, z-osu, n-carbobenzoxyoxysuccinimide, n-benzyloxycarbonyloxy-succinimide, cbzosu, carbobenzoxy succinimide | |
| MJSHDCCLFGOEIK-UHFFFAOYSA-N | |
| benzyl (2,5-dioxopyrrolidin-1-yl) carbonate | |
| 83172 | |
| 98% | |
| N-(Benzyloxycarbonyloxy)succinimide |
| 97.5 | |
| 77°C to 82°C | |
| 0.25% max. (60°C, 2 hrs, 1mm Hg) | |
| 97.5% min. (HPLC) | |
| C12H11NO5 | |
| 100 g | |
| (10% in acetone) Clear colorless to almost colorless | |
| C1CC(=O)N(C1=O)OC(=O)OCC2=CC=CC=C2 | |
| 249.22 | |
| 249.22 | |
| Fine Crystalline Powder |
Safety and Handling
EINECSNumber : 236-075-3
RUO – Research Use Only