missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyloxycarbonylglycine methyl ester, 98%

Catalog No. p-7041622
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6285806 5 g
AAH6285814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6285806 Supplier Thermo Scientific Chemicals Supplier No. H6285806

CAS: 1212-53-9 | C11H13NO4 | 223.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1212-53-9
Molecular Formula C11H13NO4
Molecular Weight (g/mol) 223.23
MDL Number MFCD00043631
InChI Key DZYBBBYFLOPVOL-UHFFFAOYSA-N
Synonym z-gly-ome, z-glycine methyl ester, methyl 2-benzyloxy carbonyl amino acetate, carbobenzyloxyglycine methyl ester, n-cbz-glycine methyl ester, methyl n-benzyloxycarbonylglycinate, cbz-glycine methyl ester, carbobenzoxyglycine methyl ester, methyl 2-benzyloxycarbonylamino acetate, glycine, n-phenylmethoxy carbonyl-, methyl ester
PubChem CID 71033
SMILES COC(=O)CNC(=O)OCC1=CC=CC=C1

Specifications

Melting Point 22°C to 26°C
Boiling Point 147°C to 151°C
Linear Formula C6H5CH2OCONHCH2CO2CH3
Quantity 5 g
IUPAC Name methyl 2-(phenylmethoxycarbonylamino)acetate
Formula Weight 223.23
Percent Purity 98%
Chemical Name or Material N-Benzyloxycarbonylglycine methyl ester
EINECSNumber 214-922-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only