missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Benzyloxycarbonyl-O-tert-butyl-L-tyrosine, 98%
CAS: 5545-54-0 | C21H25NO5 | 371.43 g/mol
$79.44 - $898.53
Chemical Identifiers
| CAS | 5545-54-0 |
|---|---|
| Molecular Formula | C21H25NO5 |
| Molecular Weight (g/mol) | 371.43 |
| MDL Number | MFCD00065701 |
| InChI Key | YKVBQSGNGCKQSV-UHFFFAOYNA-N |
| Synonym | z-tyr tbu-oh, s-2-benzyloxy carbonyl amino-3-4-tert-butoxy phenyl propanoic acid, n-cbz-o-tert-butyl-l-tyrosine, l-tyrosine, o-1,1-dimethylethyl-n-phenylmethoxy carbonyl, 2s-2-benzyloxy carbonyl amino-3-4-tert-butoxy phenyl propanoic acid, z-tyr but-oh, z-l-tyr tbu-oh, z-tyr t-bu-oh, n-benzyloxycarbonyl-o-t-butyl-l-tyrosine, o-tert-butyl-n-benzyloxycarbonyl tyrosine |
| PubChem CID | 978520 |
| IUPAC Name | 2-{[(benzyloxy)carbonyl]amino}-3-[4-(tert-butoxy)phenyl]propanoic acid |
| SMILES | CC(C)(C)OC1=CC=C(CC(NC(=O)OCC2=CC=CC=C2)C(O)=O)C=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6676206
|
Thermo Scientific Chemicals
H6676206 |
5 g |
Each for $79.44
|
|
|||||
|
AAH6676222
|
Thermo Scientific Chemicals
H6676222 |
100 g |
Each for $898.53
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 5545-54-0 | |
| 371.43 | |
| YKVBQSGNGCKQSV-UHFFFAOYNA-N | |
| 978520 | |
| CC(C)(C)OC1=CC=C(CC(NC(=O)OCC2=CC=CC=C2)C(O)=O)C=C1 |
| C21H25NO5 | |
| MFCD00065701 | |
| z-tyr tbu-oh, s-2-benzyloxy carbonyl amino-3-4-tert-butoxy phenyl propanoic acid, n-cbz-o-tert-butyl-l-tyrosine, l-tyrosine, o-1,1-dimethylethyl-n-phenylmethoxy carbonyl, 2s-2-benzyloxy carbonyl amino-3-4-tert-butoxy phenyl propanoic acid, z-tyr but-oh, z-l-tyr tbu-oh, z-tyr t-bu-oh, n-benzyloxycarbonyl-o-t-butyl-l-tyrosine, o-tert-butyl-n-benzyloxycarbonyl tyrosine | |
| 2-{[(benzyloxy)carbonyl]amino}-3-[4-(tert-butoxy)phenyl]propanoic acid |
Specifications
| 5545-54-0 | |
| C21H25NO5 | |
| 2781067 | |
| YKVBQSGNGCKQSV-UHFFFAOYNA-N | |
| 2-{[(benzyloxy)carbonyl]amino}-3-[4-(tert-butoxy)phenyl]propanoic acid | |
| 371.43 | |
| 371.43 | |
| Powder |
| White | |
| MFCD00065701 | |
| z-tyr tbu-oh, s-2-benzyloxy carbonyl amino-3-4-tert-butoxy phenyl propanoic acid, n-cbz-o-tert-butyl-l-tyrosine, l-tyrosine, o-1,1-dimethylethyl-n-phenylmethoxy carbonyl, 2s-2-benzyloxy carbonyl amino-3-4-tert-butoxy phenyl propanoic acid, z-tyr but-oh, z-l-tyr tbu-oh, z-tyr t-bu-oh, n-benzyloxycarbonyl-o-t-butyl-l-tyrosine, o-tert-butyl-n-benzyloxycarbonyl tyrosine | |
| CC(C)(C)OC1=CC=C(CC(NC(=O)OCC2=CC=CC=C2)C(O)=O)C=C1 | |
| 5 g | |
| 978520 | |
| 98% | |
| N-Benzyloxycarbonyl-O-tert-butyl-L-tyrosine |
Safety and Handling
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only