accessibility menu, dialog, popup

UserName

CAS RN 5545-54-0

O(S)OOHHNOOH3CCH3CH3

CAS RN 5545-54-0

IUPAC Name: 2-{[(benzyloxy)carbonyl]amino}-3-[4-(tert-butoxy)phenyl]propanoic acid
Synonyms: 2-{[(benzyloxy)carbonyl]amino}-3-[4-(tert-butoxy)phenyl]propanoic acid
Molecular Weight (g/mol): 371.43
Molecular Formula: C21H25NO5
InChi Key: YKVBQSGNGCKQSV-UHFFFAOYNA-N
SMILES: CC(C)(C)OC1=CC=C(CC(NC(=O)OCC2=CC=CC=C2)C(O)=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)