missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyloxycarbonyl-L-valine, 99%

Catalog No. AAA1358322
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1358322 100 g
AAA1358314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1358322 Supplier Thermo Scientific Chemicals Supplier No. A1358322
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1149-26-4 | C13H17NO4 | 251.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1149-26-4
Molecular Formula C13H17NO4
Molecular Weight (g/mol) 251.28
MDL Number MFCD00008922
InChI Key CANZBRDGRHNSGZ-NSHDSACASA-N
Synonym n-carbobenzyloxy-l-valine, n-cbz-l-valine, z-val-oh, cbz-l-valine, n-benzyloxycarbonyl-l-valine, n-carbobenzoxy-l-valine, z-l-val-oh, carbobenzoxy-l-valine, z-valine, carbobenzyloxy-l-valine
PubChem CID 726987
IUPAC Name (2S)-3-methyl-2-(phenylmethoxycarbonylamino)butanoic acid
SMILES CC(C)[C@H](NC(=O)OCC1=CC=CC=C1)C(O)=O

Specifications

Melting Point 60°C
Quantity 100 g
Beilstein 2056617
Formula Weight 251.28
Percent Purity 99%
Chemical Name or Material N-Benzyloxycarbonyl-L-valine
EINECSNumber 214-562-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only