missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyloxycarbonyl-L-tyrosine methyl ester, 98%

CAS: 13512-31-7 | C18H19NO5 | 329.35 g/mol

Supplier:  Thermo Scientific Chemicals H6311314

 View more versions of this product

Catalog No. AAH6311314


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

93°C to 97°C (decomposition)
13512-31-7
MFCD00038784
SLLMDHBKALJDBW-XISACWJONA-N
methyl (2S)-3-(4-hydroxyphenyl)-2-(phenylmethoxycarbonylamino)propanoate
329.35
329.35
Solid
N-Benzyloxycarbonyl-L-tyrosine methyl ester
C18H19NO5
z-tyr-ome, z-l-tyrosine methyl ester, n-cbz-l-tyrosine methyl ester, s-methyl 2-benzyloxy carbonyl amino-3-4-hydroxyphenyl propanoate, cbz-tyr-ome, z-l-tyr-ome, z-tyr-och3, cbz-tyrosine methyl ester, n-cbz-l-tyrosinemethylester, n-benzyloxycarbonyltyrosine methyl ester
COC(=O)[C@H](CC1=CC=C(O)C=C1)NC(=O)OCC1=CC=CC=C1
25 g
7010613
98%

Chemical Identifiers

13512-31-7
329.35
SLLMDHBKALJDBW-XISACWJONA-N
7010613
C18H19NO5
MFCD00038784
z-tyr-ome, z-l-tyrosine methyl ester, n-cbz-l-tyrosine methyl ester, s-methyl 2-benzyloxy carbonyl amino-3-4-hydroxyphenyl propanoate, cbz-tyr-ome, z-l-tyr-ome, z-tyr-och3, cbz-tyrosine methyl ester, n-cbz-l-tyrosinemethylester, n-benzyloxycarbonyltyrosine methyl ester
COC(=O)[C@H](CC1=CC=C(O)C=C1)NC(=O)OCC1=CC=CC=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 426-320-4

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only