accessibility menu, dialog, popup

UserName

(S)OHOHNOOOCH3

CAS RN 13512-31-7

IUPAC Name: methyl (2S)-2-{[(benzyloxy)carbonyl]amino}-3-(4-hydroxyphenyl)propanoate
Synonyms: methyl (2S)-2-{[(benzyloxy)carbonyl]amino}-3-(4-hydroxyphenyl)propanoate
Molecular Weight (g/mol): 329.35
Molecular Formula: C18H19NO5
InChi Key: SLLMDHBKALJDBW-XISACWJONA-N
SMILES: COC(=O)[C@H](CC1=CC=C(O)C=C1)NC(=O)OCC1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)