missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyloxycarbonyl-L-tyrosine methyl ester, 98%

Catalog No. AAH6311306
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6311306 5 g
AAH6311314 25 g
AAH6311322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6311306 Supplier Thermo Scientific Chemicals Supplier No. H6311306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 13512-31-7 | C18H19NO5 | 329.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13512-31-7
Molecular Formula C18H19NO5
Molecular Weight (g/mol) 329.35
MDL Number MFCD00038784
InChI Key SLLMDHBKALJDBW-XISACWJONA-N
Synonym z-tyr-ome, z-l-tyrosine methyl ester, n-cbz-l-tyrosine methyl ester, s-methyl 2-benzyloxy carbonyl amino-3-4-hydroxyphenyl propanoate, cbz-tyr-ome, z-l-tyr-ome, z-tyr-och3, cbz-tyrosine methyl ester, n-cbz-l-tyrosinemethylester, n-benzyloxycarbonyltyrosine methyl ester
PubChem CID 7010613
SMILES COC(=O)[C@H](CC1=CC=C(O)C=C1)NC(=O)OCC1=CC=CC=C1

Specifications

Melting Point 93°C to 97°C (decomposition)
IUPAC Name methyl 2-{[(benzyloxy)carbonyl]amino}-3-(4-hydroxyphenyl)propanoate
Quantity 5 g
Formula Weight 329.35
Percent Purity 98%
Physical Form Solid
Chemical Name or Material N-Benzyloxycarbonyl-L-tyrosine methyl ester
EINECSNumber 426-320-4
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only