missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyloxycarbonyl-L-threonine, 99%

Catalog No. AAA1756506
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1756506 5 g
AAA1756514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1756506 Supplier Thermo Scientific Chemicals Supplier No. A1756506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 19728-63-3 | C12H15NO5 | 253.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 19728-63-3
Molecular Formula C12H15NO5
Molecular Weight (g/mol) 253.25
MDL Number MFCD00065948
InChI Key IPJUIRDNBFZGQN-SCZZXKLOSA-N
Synonym z-thr-oh, n-cbz-l-threonine, z-l-threonine, z-l-thr-oh, n-benzyloxycarbonyl-l-threonine, cbz-l-threonine, n-carbobenzyloxy-l-threonine, l-threonine, n-phenylmethoxy carbonyl, benzyloxycarbonyl-l-threonine, n-benzyloxy carbonyl-l-threonine
PubChem CID 88217
IUPAC Name (2S,3R)-3-hydroxy-2-(phenylmethoxycarbonylamino)butanoic acid
SMILES C[C@@H](O)[C@H](NC(=O)OCC1=CC=CC=C1)C(O)=O

Specifications

Melting Point 103°C to 105°C
Quantity 5 g
Beilstein 2335409
Optical Rotation −4.5° (c=4 in Acetic Acid)
Formula Weight 253.26
Percent Purity 99%
Chemical Name or Material N-Benzyloxycarbonyl-L-threonine
EINECSNumber 243-258-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only