accessibility menu, dialog, popup

UserName

H3C(R)(S)OHOHNOHOO

CAS RN 19728-63-3

IUPAC Name: (2S,3R)-2-{[(benzyloxy)carbonyl]amino}-3-hydroxybutanoic acid
Synonyms: (2S,3R)-2-{[(benzyloxy)carbonyl]amino}-3-hydroxybutanoic acid
Molecular Weight (g/mol): 253.25
Molecular Formula: C12H15NO5
InChi Key: IPJUIRDNBFZGQN-SCZZXKLOSA-N
SMILES: C[C@@H](O)[C@H](NC(=O)OCC1=CC=CC=C1)C(O)=O
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)