missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyloxycarbonyl-L-serine, 99%

Catalog No. p-7024423
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
25 g
5 g
Catalog No. Quantity
AAA1905006 5 g
AAA1905014 25 g
AAA1905022 100 g
3 options

Catalog No. AAA1905006

Supplier: Thermo Scientific Chemicals A1905006

Only null left

CAS: 1145-80-8 | C11H13NO5 | 239.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1145-80-8
Molecular Formula C11H13NO5
Molecular Weight (g/mol) 239.23
MDL Number MFCD00002662
InChI Key GNIDSOFZAKMQAO-UHFFFAOYNA-N
Synonym z-ser-oh, n-cbz-l-serine, benzyloxycarbonylserine, z-l-serine, carbobenzyloxy-l-serine, n-carbobenzyloxy-l-serine, cbz-l-ser-oh, n-carbobenzoxy-l-serine, n-benzyloxycarbonyl-l-serine, cbz-l-serine
PubChem CID 100310
IUPAC Name (2S)-3-hydroxy-2-(phenylmethoxycarbonylamino)propanoic acid
SMILES OCC(NC(=O)OCC1=CC=CC=C1)C(O)=O

Specifications

Melting Point 117°C to 120°C
Quantity 5 g
Beilstein 2058314
Optical Rotation +6° (c=7 in Acetic Acid)
Formula Weight 239.23
Percent Purity 99%
Chemical Name or Material N-Benzyloxycarbonyl-L-serine
EINECSNumber 214-546-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only