missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyloxycarbonyl-L-serine, 99%

Catalog No. AAA1905006
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1905006 5 g
AAA1905014 25 g
AAA1905022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1905006 Supplier Thermo Scientific Chemicals Supplier No. A1905006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1145-80-8 | C11H13NO5 | 239.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1145-80-8
Molecular Formula C11H13NO5
Molecular Weight (g/mol) 239.23
MDL Number MFCD00002662
InChI Key GNIDSOFZAKMQAO-UHFFFAOYNA-N
Synonym z-ser-oh, n-cbz-l-serine, benzyloxycarbonylserine, z-l-serine, carbobenzyloxy-l-serine, n-carbobenzyloxy-l-serine, cbz-l-ser-oh, n-carbobenzoxy-l-serine, n-benzyloxycarbonyl-l-serine, cbz-l-serine
PubChem CID 100310
IUPAC Name (2S)-3-hydroxy-2-(phenylmethoxycarbonylamino)propanoic acid
SMILES OCC(NC(=O)OCC1=CC=CC=C1)C(O)=O

Specifications

Melting Point 117°C to 120°C
Quantity 5 g
Beilstein 2058314
Optical Rotation +6° (c=7 in Acetic Acid)
Formula Weight 239.23
Percent Purity 99%
Chemical Name or Material N-Benzyloxycarbonyl-L-serine
EINECSNumber 214-546-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only