missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyloxycarbonyl-L-leucine N-succinimidyl ester, 95%

Catalog No. p-7056558
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6554606 5 g
AAH6554614 25 g
AAH6554622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6554606 Supplier Thermo Scientific Chemicals Supplier No. H6554606

CAS: 3397-35-1 | C18H22N2O6 | 362.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3397-35-1
Molecular Formula C18H22N2O6
Molecular Weight (g/mol) 362.38
MDL Number MFCD00042759
InChI Key YHZUOMRURVTBMO-YQTOOIBONA-N
Synonym z-leu-osu, n-carbobenzoxy-l-leucine n-succinimidyl ester, s-2,5-dioxopyrrolidin-1-yl 2-benzyloxy carbonyl amino-4-methylpentanoate, 2,5-dioxopyrrolidin-1-yl 2s-2-benzyloxy carbonyl amino-4-methylpentanoate, 2,5-dioxoazolidinyl 2s-4-methyl-2-phenylmethoxy carbonylamino pentanoate, z-l-leucine hydroxysuccinimide ester, n-cbz-l-leucine n-succinimidyl ester, z-l-leucine n-hydroxysuccinimide ester, n-cbz-l-leucine n-hydroxysuccinimide ester, z-l-leucine hydroxysuccinimide ester gr
PubChem CID 11089751
IUPAC Name 2,5-dioxopyrrolidin-1-yl (2S)-2-{[(benzyloxy)carbonyl]amino}-4-methylpentanoate
SMILES CC(C)C[C@H](NC(=O)OCC1=CC=CC=C1)C(=O)ON1C(=O)CCC1=O

Specifications

Melting Point 116°C to 120°C
Color White
Beilstein 1555663
Quantity 5 g
Formula Weight 362.38
Percent Purity 95%
Physical Form Solid
Chemical Name or Material N-Benzyloxycarbonyl-L-leucine N-succinimidyl ester
EINECSNumber 222-256-4
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only