accessibility menu, dialog, popup

UserName

CH3CH3(S)NHOOOONOO

CAS RN 3397-35-1

IUPAC Name: 2,5-dioxopyrrolidin-1-yl (2S)-2-{[(benzyloxy)carbonyl]amino}-4-methylpentanoate
Synonyms: 2,5-dioxopyrrolidin-1-yl (2S)-2-{[(benzyloxy)carbonyl]amino}-4-methylpentanoate
Molecular Weight (g/mol): 362.38
Molecular Formula: C18H22N2O6
InChi Key: YHZUOMRURVTBMO-YQTOOIBONA-N
SMILES: CC(C)C[C@H](NC(=O)OCC1=CC=CC=C1)C(=O)ON1C(=O)CCC1=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)