missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyloxycarbonyl-L-glutamic acid gamma-tert-butyl ester, 98+%, Thermo Scientific™

Chemical Identifiers

CAS 3886-08-6
Molecular Formula C17H23NO6
Molecular Weight (g/mol) 337.37
MDL Number MFCD00038274
InChI Key GLMODRZPPBZPPB-ZDUSSCGKSA-M
IUPAC Name (2S)-2-{[(benzyloxy)carbonyl]amino}-5-(tert-butoxy)-5-oxopentanoate
SMILES CC(C)(C)OC(=O)CC[C@H](NC(=O)OCC1=CC=CC=C1)C([O-])=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC426150050
View Documents
Thermo Scientific Chemicals
426150050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

3886-08-6
337.37
GLMODRZPPBZPPB-ZDUSSCGKSA-M
CC(C)(C)OC(=O)CC[C@H](NC(=O)OCC1=CC=CC=C1)C([O-])=O
C17H23NO6
MFCD00038274
(2S)-2-{[(benzyloxy)carbonyl]amino}-5-(tert-butoxy)-5-oxopentanoate

Specifications

3886-08-6
100.0
Glass bottle
MFCD00038274
Solubility in water: insoluble
CC(C)(C)OC(=O)CC[C@H](NC(=O)OCC1=CC=CC=C1)C([O-])=O
337.37
98+%
98.0
97.5% min. (HPLC)
C17H23NO6
5g
GLMODRZPPBZPPB-ZDUSSCGKSA-M
(2S)-2-{[(benzyloxy)carbonyl]amino}-5-(tert-butoxy)-5-oxopentanoate
337.37
N-Benzyloxycarbonyl-L-glutamic acid γ-tert-butyl ester
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 223-421-3

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only