missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Benzyloxycarbonyl-L-glutamic acid gamma-tert-butyl ester, 98+%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 426150050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| N-Benzyloxycarbonyl-L-glutamic acid γ-tert-butyl ester | |
| 98.0 | |
| 97.5% min. (HPLC) | |
| C17H23NO6 | |
| 5g | |
| GLMODRZPPBZPPB-ZDUSSCGKSA-M | |
| (2S)-2-{[(benzyloxy)carbonyl]amino}-5-(tert-butoxy)-5-oxopentanoate | |
| 337.37 |
| 3886-08-6 | |
| 100.0 | |
| Glass bottle | |
| MFCD00038274 | |
| Solubility in water: insoluble | |
| CC(C)(C)OC(=O)CC[C@H](NC(=O)OCC1=CC=CC=C1)C([O-])=O | |
| 337.37 | |
| 98+% |
Chemical Identifiers
| 3886-08-6 | |
| 337.37 | |
| GLMODRZPPBZPPB-ZDUSSCGKSA-M | |
| CC(C)(C)OC(=O)CC[C@H](NC(=O)OCC1=CC=CC=C1)C([O-])=O |
| C17H23NO6 | |
| MFCD00038274 | |
| (2S)-2-{[(benzyloxy)carbonyl]amino}-5-(tert-butoxy)-5-oxopentanoate |
Safety and Handling
EINECSNumber : 223-421-3
RUO â Research Use Only