missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Benzyloxycarbonyl-L-glutamic acid 1-tert-butyl ester, 95%
CAS: 5891-45-2 | C17H23NO6 | 337.37 g/mol
$137.34 - $609.03
Chemical Identifiers
| CAS | 5891-45-2 |
|---|---|
| Molecular Formula | C17H23NO6 |
| Molecular Weight (g/mol) | 337.37 |
| MDL Number | MFCD00077016 |
| InChI Key | VJECGKAFPHEJQS-UHFFFAOYNA-N |
| Synonym | z-glu-otbu, a-tert.butyl ester, s-4-benzyloxy carbonyl amino-5-tert-butoxy-5-oxopentanoic acid, cbz-glu-otbu, z-glu-o-tbu, pubchem18088, pubchem18088, 1-tert-butyl n-cbz-l-glutamate, 1-tert-butyl n-cbz-l-glutamate, z-l-glutamic acid, z-l-glutamic acid |
| PubChem CID | 6994036 |
| IUPAC Name | 4-{[(benzyloxy)carbonyl]amino}-5-(tert-butoxy)-5-oxopentanoic acid |
| SMILES | CC(C)(C)OC(=O)C(CCC(O)=O)NC(=O)OCC1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6395403
|
Thermo Scientific Chemicals
H6395403 |
1 g |
Each for $137.34
|
|
|||||
|
AAH6395406
|
Thermo Scientific Chemicals
H6395406 |
5 g |
Each for $609.03
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 5891-45-2 | |
| 337.37 | |
| VJECGKAFPHEJQS-UHFFFAOYNA-N | |
| 6994036 | |
| CC(C)(C)OC(=O)C(CCC(O)=O)NC(=O)OCC1=CC=CC=C1 |
| C17H23NO6 | |
| MFCD00077016 | |
| z-glu-otbu, a-tert.butyl ester, s-4-benzyloxy carbonyl amino-5-tert-butoxy-5-oxopentanoic acid, cbz-glu-otbu, z-glu-o-tbu, pubchem18088, pubchem18088, 1-tert-butyl n-cbz-l-glutamate, 1-tert-butyl n-cbz-l-glutamate, z-l-glutamic acid, z-l-glutamic acid | |
| 4-{[(benzyloxy)carbonyl]amino}-5-(tert-butoxy)-5-oxopentanoic acid |
Specifications
| 5891-45-2 | |
| MFCD00077016 | |
| VJECGKAFPHEJQS-UHFFFAOYNA-N | |
| 4-{[(benzyloxy)carbonyl]amino}-5-(tert-butoxy)-5-oxopentanoic acid | |
| 337.37 | |
| 337.37 | |
| Solid |
| C17H23NO6 | |
| z-glu-otbu, a-tert.butyl ester, s-4-benzyloxy carbonyl amino-5-tert-butoxy-5-oxopentanoic acid, cbz-glu-otbu, z-glu-o-tbu, pubchem18088, pubchem18088, 1-tert-butyl n-cbz-l-glutamate, 1-tert-butyl n-cbz-l-glutamate, z-l-glutamic acid, z-l-glutamic acid | |
| CC(C)(C)OC(=O)C(CCC(O)=O)NC(=O)OCC1=CC=CC=C1 | |
| 1 g | |
| 6994036 | |
| 95% | |
| N-Benzyloxycarbonyl-L-glutamic acid 1-tert-butyl ester |
Safety and Handling
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only