missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Benzyloxycarbonyl-L-glutamic acid 1-tert-butyl ester, 95%
CAS: 5891-45-2 | C17H23NO6 | 337.37 g/mol
Supplier: Thermo Scientific Chemicals H6395406
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| N-Benzyloxycarbonyl-L-glutamic acid 1-tert-butyl ester | |
| C17H23NO6 | |
| z-glu-otbu, a-tert.butyl ester, s-4-benzyloxycarbonylamino-5-tert-butoxy-5-oxopentanoic acid, s-4-benzyloxycarbonylamino-5-tert-butoxy-5-oxopentanoic acid, cbz-glu-otbu, pubchem18088, pubchem18088, 1-tert-butyl n-cbz-l-glutamate, l-glutamic acid, n-phenylmethoxy carbonyl-, 1-1,1-dimethylethyl ester, z-l-glutamic acid, z-l-glutamic acid | |
| CC(C)(C)OC(=O)C(CCC(O)=O)NC(=O)OCC1=CC=CC=C1 | |
| 5 g | |
| 6994036 | |
| 95% |
| 5891-45-2 | |
| MFCD00077016 | |
| VJECGKAFPHEJQS-UHFFFAOYNA-N | |
| 4-{[(benzyloxy)carbonyl]amino}-5-(tert-butoxy)-5-oxopentanoic acid | |
| 337.37 | |
| 337.37 | |
| Solid |
Chemical Identifiers
| 5891-45-2 | |
| 337.37 | |
| VJECGKAFPHEJQS-UHFFFAOYNA-N | |
| 6994036 | |
| CC(C)(C)OC(=O)C(CCC(O)=O)NC(=O)OCC1=CC=CC=C1 |
| C17H23NO6 | |
| MFCD00077016 | |
| z-glu-otbu, a-tert.butyl ester, s-4-benzyloxycarbonylamino-5-tert-butoxy-5-oxopentanoic acid, s-4-benzyloxycarbonylamino-5-tert-butoxy-5-oxopentanoic acid, cbz-glu-otbu, pubchem18088, pubchem18088, 1-tert-butyl n-cbz-l-glutamate, l-glutamic acid, n-phenylmethoxy carbonyl-, 1-1,1-dimethylethyl ester, z-l-glutamic acid, z-l-glutamic acid | |
| 4-{[(benzyloxy)carbonyl]amino}-5-(tert-butoxy)-5-oxopentanoic acid |
Safety and Handling
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only