missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Benzyloxycarbonyl-L-aspartic acid 1-benzyl ester, 95%
CAS: 4779-31-1 | C19H19NO6 | 357.36 g/mol
Supplier: Thermo Scientific Chemicals H6230606
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| N-Benzyloxycarbonyl-L-aspartic acid 1-benzyl ester | |
| C19H19NO6 | |
| 5 g | |
| UYOZWZKJQRBZRH-UHFFFAOYNA-N | |
| 4-(benzyloxy)-3-{[(benzyloxy)carbonyl]amino}-4-oxobutanoic acid | |
| 6994509 | |
| 95% |
| 4779-31-1 | |
| MFCD00038594 | |
| z-asp-obzl, n-carbobenzyloxy-l-aspartic acid 1-benzyl ester, 1-benzyl n-carbobenzyloxy-l-aspartate, 3s-4-benzyloxy-3-benzyloxy carbonyl amino-4-oxobutanoic acid, 3s-4-oxo-4-phenylmethoxy-3-phenylmethoxycarbonylamino butanoic acid, z-asp-obn, cbz-l-asp-obn, benzyloxycarbonyl-l-aspartic-benzyl ester, n-benzyloxycarbonyl-l-aspartic acid 1-benzyl ester, 2-benzyl-oxycarbonylamino-succinic acid 1-benzyl ester | |
| OC(=O)CC(NC(=O)OCC1=CC=CC=C1)C(=O)OCC1=CC=CC=C1 | |
| 357.36 | |
| 357.36 |
Chemical Identifiers
| 4779-31-1 | |
| 357.36 | |
| UYOZWZKJQRBZRH-UHFFFAOYNA-N | |
| 6994509 | |
| OC(=O)CC(NC(=O)OCC1=CC=CC=C1)C(=O)OCC1=CC=CC=C1 |
| C19H19NO6 | |
| MFCD00038594 | |
| z-asp-obzl, n-carbobenzyloxy-l-aspartic acid 1-benzyl ester, 1-benzyl n-carbobenzyloxy-l-aspartate, 3s-4-benzyloxy-3-benzyloxy carbonyl amino-4-oxobutanoic acid, 3s-4-oxo-4-phenylmethoxy-3-phenylmethoxycarbonylamino butanoic acid, z-asp-obn, cbz-l-asp-obn, benzyloxycarbonyl-l-aspartic-benzyl ester, n-benzyloxycarbonyl-l-aspartic acid 1-benzyl ester, 2-benzyl-oxycarbonylamino-succinic acid 1-benzyl ester | |
| 4-(benzyloxy)-3-{[(benzyloxy)carbonyl]amino}-4-oxobutanoic acid |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only