missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyloxycarbonyl-L-aspartic acid 1-benzyl ester, 95%

Catalog No. p-7041489
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6230603 1 g
AAH6230606 5 g
AAH6230614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6230603 Supplier Thermo Scientific Chemicals Supplier No. H6230603
Only null left

CAS: 4779-31-1 | C19H19NO6 | 357.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4779-31-1
Molecular Formula C19H19NO6
Molecular Weight (g/mol) 357.36
MDL Number MFCD00038594
InChI Key UYOZWZKJQRBZRH-UHFFFAOYNA-N
Synonym z-asp-obzl, n-carbobenzyloxy-l-aspartic acid 1-benzyl ester, 1-benzyl n-carbobenzyloxy-l-aspartate, 3s-4-benzyloxy-3-benzyloxy carbonyl amino-4-oxobutanoic acid, 3s-4-oxo-4-phenylmethoxy-3-phenylmethoxycarbonylamino butanoic acid, z-asp-obn, cbz-l-asp-obn, benzyloxycarbonyl-l-aspartic-benzyl ester, n-benzyloxycarbonyl-l-aspartic acid 1-benzyl ester, 2-benzyl-oxycarbonylamino-succinic acid 1-benzyl ester
PubChem CID 6994509
IUPAC Name 4-(benzyloxy)-3-{[(benzyloxy)carbonyl]amino}-4-oxobutanoic acid
SMILES OC(=O)CC(NC(=O)OCC1=CC=CC=C1)C(=O)OCC1=CC=CC=C1

Specifications

Quantity 1 g
Formula Weight 357.36
Percent Purity 95%
Chemical Name or Material N-Benzyloxycarbonyl-L-aspartic acid 1-benzyl ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only