missing translation for 'onlineSavingsMsg'
Learn More

N-(Benzyloxycarbonyl)hydroxylamine, 98+%

CAS: 3426-71-9 | C8H9NO3 | 167.16 g/mol

Supplier:  Thermo Scientific Chemicals L1317514

 View more versions of this product

Catalog No. AAL1317514


Only null left
Explore available promotions

Description

Description

N-(Benzyloxycarbonyl)hydroxylamine reacts with cyclohepta-1,3-diene to produce 6-oxa-7-aza-bicyclo[3.2.2]non-8-ene-7-carboxylic acid benzyl ester.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-(Benzyloxycarbonyl)hydroxylamine reacts with cyclohepta-1,3-diene to produce 6-oxa-7-aza-bicyclo[3.2.2]non-8-ene-7-carboxylic acid benzyl ester.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is sensitive to moisture. Incompatible with strong oxidizing agents. Store at -20°C.
Specifications

Specifications

N-(Benzyloxycarbonyl)hydroxylamine
65°C to 70°C
MFCD00010642
1074034
benzyl hydroxycarbamate, n-benzyloxycarbonyl hydroxylamine, n-carbobenzoxyhydroxylamine, carbamic acid, hydroxy-, phenylmethyl ester, n-benzyloxycarbonylhydroxylamine, benzylhydroxycarbamate, ccris 5076, carbamic acid, hydroxy-, benzyl ester, 1-06-00-00221 beilstein handbook reference, carbamic acid, n-hydroxy-, phenylmethyl ester
ONC(=O)OCC1=CC=CC=C1
167.16
167.17
3426-71-9
C8H9NO3
25 g
Moisture sensitive
PQBSPTAPCMSZAA-UHFFFAOYSA-N
benzyl N-hydroxycarbamate
18907
≥98%

Chemical Identifiers

3426-71-9
167.16
PQBSPTAPCMSZAA-UHFFFAOYSA-N
18907
ONC(=O)OCC1=CC=CC=C1
C8H9NO3
MFCD00010642
benzyl hydroxycarbamate, n-benzyloxycarbonyl hydroxylamine, n-carbobenzoxyhydroxylamine, carbamic acid, hydroxy-, phenylmethyl ester, n-benzyloxycarbonylhydroxylamine, benzylhydroxycarbamate, ccris 5076, carbamic acid, hydroxy-, benzyl ester, 1-06-00-00221 beilstein handbook reference, carbamic acid, n-hydroxy-, phenylmethyl ester
benzyl N-hydroxycarbamate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 000-000-0

RTECSNumber : FB1600000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only