missing translation for 'onlineSavingsMsg'
Learn More

N-(Benzyloxycarbonyl)hydroxylamine, 98+%

Catalog No. p-7046119
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1317506 5 g
AAL1317514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1317506 Supplier Thermo Scientific Chemicals Supplier No. L1317506

CAS: 3426-71-9 | C8H9NO3 | 167.16 g/mol

N-(Benzyloxycarbonyl)hydroxylamine reacts with cyclohepta-1,3-diene to produce 6-oxa-7-aza-bicyclo[3.2.2]non-8-ene-7-carboxylic acid benzyl ester.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-(Benzyloxycarbonyl)hydroxylamine reacts with cyclohepta-1,3-diene to produce 6-oxa-7-aza-bicyclo[3.2.2]non-8-ene-7-carboxylic acid benzyl ester.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is sensitive to moisture. Incompatible with strong oxidizing agents. Store at -20°C.

Chemical Identifiers

CAS 3426-71-9
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.16
MDL Number MFCD00010642
InChI Key PQBSPTAPCMSZAA-UHFFFAOYSA-N
Synonym benzyl hydroxycarbamate, n-benzyloxycarbonyl hydroxylamine, n-carbobenzoxyhydroxylamine, carbamic acid, hydroxy-, phenylmethyl ester, n-benzyloxycarbonylhydroxylamine, benzylhydroxycarbamate, ccris 5076, carbamic acid, hydroxy-, benzyl ester, 1-06-00-00221 beilstein handbook reference, carbamic acid, n-hydroxy-, phenylmethyl ester
PubChem CID 18907
IUPAC Name benzyl N-hydroxycarbamate
SMILES ONC(=O)OCC1=CC=CC=C1

Specifications

Melting Point 65°C to 70°C
Quantity 5 g
Beilstein 1074034
Sensitivity Moisture sensitive
Formula Weight 167.17
Percent Purity ≥98%
Chemical Name or Material N-(Benzyloxycarbonyl)hydroxylamine
EINECSNumber 000-000-0
RTECSNumber FB1600000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only